missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxytryptamine, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC187450500

 View more versions of this product

Catalog No. AC187450500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

6-Methoxytryptamine
143.0°C to 146.0°C
99%
98.5
99%
MFCD00005663
VOCGEKMEZOPDFP-UHFFFAOYSA-N
2-(6-methoxy-1H-indol-3-yl)ethanamine
17654
99%
Glass bottle
50mg
3610-36-4
100.0
C11H14N2O
6-methoxytryptamine, 2-6-methoxy-1h-indol-3-yl ethanamine, 3-2-aminoethyl-6-methoxyindole, 2-6-methoxy-1h-indol-3-yl-ethylamine, 1h-indole-3-ethanamine, 6-methoxy, 2-2-aminoethyl-5-methoxyindole, 1h-indole-3-ethanamine,6-methoxy, 2-6-methoxy-1h-indol-3-yl ethan-1-amine, 2-6-methoxyindol-3-yl ethylamine, 6-methoxy tryptamine
COC1=CC2=C(C=C1)C(=CN2)CCN
190.246
190.24

Chemical Identifiers

3610-36-4
190.246
VOCGEKMEZOPDFP-UHFFFAOYSA-N
17654
COC1=CC2=C(C=C1)C(=CN2)CCN
C11H14N2O
MFCD00005663
6-methoxytryptamine, 2-6-methoxy-1h-indol-3-yl ethanamine, 3-2-aminoethyl-6-methoxyindole, 2-6-methoxy-1h-indol-3-yl-ethylamine, 1h-indole-3-ethanamine, 6-methoxy, 2-2-aminoethyl-5-methoxyindole, 1h-indole-3-ethanamine,6-methoxy, 2-6-methoxy-1h-indol-3-yl ethan-1-amine, 2-6-methoxyindol-3-yl ethylamine, 6-methoxy tryptamine
2-(6-methoxy-1H-indol-3-yl)ethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 222-778-2

SDS Documents