missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxysalicylic Acid, TCI America™

$32.08 - $119.17

Chemical Identifiers

CAS 3147-64-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00674090
InChI Key AAUQLHHARJUJEH-UHFFFAOYSA-N
Synonym 6-methoxysalicylic acid, 6-hydroxy-o-anisic acid, benzoic acid, 2-hydroxy-6-methoxy, 6-methoxysalicylicacid, 6-methoxysalicylsaure, pubchem18380, acmc-1cldg, 6-methoxy-salicylic acid, 2-carboxy-3-methoxyphenol, 2-carboxy-3-hydroxyanisole
PubChem CID 591524
IUPAC Name 2-hydroxy-6-methoxybenzoic acid
SMILES COC1=CC=CC(=C1C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19411G View Documents TCI America
M19411G
1g Each for $32.08
 
M19415G View Documents TCI America
M19415G
5g Each for $119.17
 
Specifications

Chemical Identifiers

3147-64-6
168.148
AAUQLHHARJUJEH-UHFFFAOYSA-N
591524
COC1=CC=CC(=C1C(=O)O)O
C8H8O4
MFCD00674090
6-methoxysalicylic acid, 6-hydroxy-o-anisic acid, benzoic acid, 2-hydroxy-6-methoxy, 6-methoxysalicylicacid, 6-methoxysalicylsaure, pubchem18380, acmc-1cldg, 6-methoxy-salicylic acid, 2-carboxy-3-methoxyphenol, 2-carboxy-3-hydroxyanisole
2-hydroxy-6-methoxybenzoic acid

Specifications

White
1g
C8H8O4
6-methoxysalicylic acid, 6-hydroxy-o-anisic acid, benzoic acid, 2-hydroxy-6-methoxy, 6-methoxysalicylicacid, 6-methoxysalicylsaure, pubchem18380, acmc-1cldg, 6-methoxy-salicylic acid, 2-carboxy-3-methoxyphenol, 2-carboxy-3-hydroxyanisole
COC1=CC=CC(=C1C(=O)O)O
168.148
168.15
≥98.0% (GC,T)
137°C
3147-64-6
MFCD00674090
AAUQLHHARJUJEH-UHFFFAOYSA-N
2-hydroxy-6-methoxybenzoic acid
591524
Crystal-Powder at 20°C
6-Methoxysalicylic Acid
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1654

TSCA : No