missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxysalicylic acid, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 437670100

 View more versions of this product

Catalog No. AC437670100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

6-Methoxysalicylic acid
97.5
White
97.5% min. (HPLC)
Authentic
2-carboxy-3-hydroxyanisole, 2-carboxy-3-methoxyphenol, 6-hydroxy-o-anisic acid, 6-methoxy-salicylic acid, 6-methoxysalicylic acid, 6-methoxysalicylicacid, 6-methoxysalicylsaure, acmc-1cldg, benzoic acid, 2-hydroxy-6-methoxy, pubchem18380
COC1=CC=CC(=C1C(=O)O)O
168.15
168.15
98%
3147-64-6
100.0
135.9°C to 136.4°C
C8H8O4
10g
AAUQLHHARJUJEH-UHFFFAOYSA-N
2-hydroxy-6-methoxybenzoic acid
591524
Powder

Chemical Identifiers

3147-64-6
168.15
2-carboxy-3-hydroxyanisole, 2-carboxy-3-methoxyphenol, 6-hydroxy-o-anisic acid, 6-methoxy-salicylic acid, 6-methoxysalicylic acid, 6-methoxysalicylicacid, 6-methoxysalicylsaure, acmc-1cldg, benzoic acid, 2-hydroxy-6-methoxy, pubchem18380
2-hydroxy-6-methoxybenzoic acid
C8H8O4
AAUQLHHARJUJEH-UHFFFAOYSA-N
591524
COC1=CC=CC(=C1C(=O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.

GHS Signal Word: Warning

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only