missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxysalicylic acid, 98%

$18.96 - $248.28

Chemical Identifiers

CAS 3147-64-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00674090
InChI Key AAUQLHHARJUJEH-UHFFFAOYSA-N
Synonym 6-methoxysalicylic acid, 6-hydroxy-o-anisic acid, benzoic acid, 2-hydroxy-6-methoxy, 6-methoxysalicylicacid, 6-methoxysalicylsaure, pubchem18380, acmc-1cldg, 6-methoxy-salicylic acid, 2-carboxy-3-methoxyphenol, 2-carboxy-3-hydroxyanisole
PubChem CID 591524
IUPAC Name 2-hydroxy-6-methoxybenzoic acid
SMILES COC1=CC=CC(=C1C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6039702 View Documents Alfa Aesar™
H6039702
0.5g Each for $18.96
 
AAH6039709 View Documents Alfa Aesar™
H6039709
10g Each for $248.28
 
Specifications

Chemical Identifiers

3147-64-6
168.148
AAUQLHHARJUJEH-UHFFFAOYSA-N
591524
COC1=CC=CC(=C1C(=O)O)O
C8H8O4
MFCD00674090
6-methoxysalicylic acid, 6-hydroxy-o-anisic acid, benzoic acid, 2-hydroxy-6-methoxy, 6-methoxysalicylicacid, 6-methoxysalicylsaure, pubchem18380, acmc-1cldg, 6-methoxy-salicylic acid, 2-carboxy-3-methoxyphenol, 2-carboxy-3-hydroxyanisole
2-hydroxy-6-methoxybenzoic acid

Specifications

134°C to 138°C
3147-64-6
MFCD00674090
AAUQLHHARJUJEH-UHFFFAOYSA-N
2-hydroxy-6-methoxybenzoic acid
591524
98%
0.5g
C8H8O4
6-methoxysalicylic acid, 6-hydroxy-o-anisic acid, benzoic acid, 2-hydroxy-6-methoxy, 6-methoxysalicylicacid, 6-methoxysalicylsaure, pubchem18380, acmc-1cldg, 6-methoxy-salicylic acid, 2-carboxy-3-methoxyphenol, 2-carboxy-3-hydroxyanisole
COC1=CC=CC(=C1C(=O)O)O
168.148
168.15
6-Methoxysalicylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning