Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxyquinoxaline, 98%

Chemical Identifiers

Chemical Name or Material 6-Methoxyquinoxaline
CAS 6639-82-3
Formula Weight 160.17
MDL Number MFCD00666601
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6339603

 
alfa aesar™
H6339603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6339606

 
alfa aesar™
H6339606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63396MD

 
alfa aesar™
H63396MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.17
MDL Number MFCD00666601
Assay 98%
Storage Note 1 2° to 8°C
CAS 6639-82-3
Chemical Name or Material 6-Methoxyquinoxaline
Quantity 1g
Physical Form Solid
Safety and Handling

2° to 8°C