missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxyquinoxaline, 98%

Manufacturer:  Alfa Aesar™ H63396MD

 View more versions of this product

Catalog No. AAH63396MD



Specifications

Specifications

6-Methoxyquinoxaline
6639-82-3
MFCD00666601
BURIFIXTNVTJJN-UHFFFAOYSA-N
6-methoxyquinoxaline
241501
Solid
250mg
C9H8N2O
quinoxaline, 6-methoxy, 6-methoxychinoxalin, ksc354o1h
COC1=CC2=NC=CN=C2C=C1
160.176
160.17
98%

Chemical Identifiers

6639-82-3
160.176
BURIFIXTNVTJJN-UHFFFAOYSA-N
241501
COC1=CC2=NC=CN=C2C=C1
C9H8N2O
MFCD00666601
quinoxaline, 6-methoxy, 6-methoxychinoxalin, ksc354o1h
6-methoxyquinoxaline
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents