missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxyquinoline, TCI America™

Manufacturer:  TCI America M089125G

 View more versions of this product

Catalog No. M089125G



Specifications

Specifications

6-Methoxyquinoline
White-Yellow
5263-87-6
MFCD00006800
HFDLDPJYCIEXJP-UHFFFAOYSA-N
6-methoxyquinoline
14860
159.19
≥97.0% (GC,T)
284°C
25g
C10H9NO
quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q
COC1=CC2=C(C=C1)N=CC=C2
159.188
CHEBI:72822
Crystal-Lump at 20°C

Chemical Identifiers

5263-87-6
159.188
HFDLDPJYCIEXJP-UHFFFAOYSA-N
14860
6-methoxyquinoline
C10H9NO
MFCD00006800
quinoline, 6-methoxy, 6-methoxy-quinoline, methyl 6-quinolyl ether, p-quinanisole, unii-5s1u3125ad, chembl15200, 6-methoxy quinoline, pubchem5917, 6-methoxyquinoline, acmc-209l0q
CHEBI:72822
COC1=CC2=C(C=C1)N=CC=C2
Safety and Handling

Safety and Handling

TSCA : Yes

Documents