Please login to your online account to display your discounted pricing

6-Methoxyquinaldine, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC146980100

 View more versions of this product

Catalog No. AC146980100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fluffy Crystalline Powder
Assay ≥96.0%
Formula Weight 173.21
Quantity 10g
Color Beige to gray
Melting Point 61° to 65°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 6-Methoxyquinaldine
Assay Percent Range 97%
Molecular Formula C11H11NO
MDL Number MFCD00006761
CAS 1078-28-0
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 6-Methoxyquinaldine
CAS 1078-28-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H11NO
Formula Weight 173.21
MDL Number MFCD00006761