missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
6-Methoxyquinaldine, 97%, ACROS Organics™
Chemical Identifiers
CAS |
1078-28-0 |
Molecular Formula |
C11H11NO |
Molecular Weight (g/mol) |
173.21 |
MDL Number |
MFCD00006761 |
InChI Key |
NAGJQQFMJKMXJQ-UHFFFAOYSA-N |
Synonym |
2-methy-6-methoxyquinolinel, 2-methyl-6-methoxyquinoline, 6-methoxy-2-methyl-quinoline, 6-methoxyquinaldine, acmc-2098xp, pubchem5910, quinaldine, 6-methoxy, quinoline, 6-methoxy-2-methyl, quinoline,6-methoxy-2-methyl |
PubChem CID |
70648 |
IUPAC Name |
6-methoxy-2-methylquinoline |
SMILES |
CC1=NC2=C(C=C1)C=C(C=C2)OC |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
AC146980010
SDS
View Documents
|
Thermo Fisher Scientific
146980010
|
1g |
Glass bottle |
N/A
|
N/A
|
|
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
|
|
Description
Specifications
Chemical Identifiers
|
1078-28-0 |
|
173.21 |
|
NAGJQQFMJKMXJQ-UHFFFAOYSA-N |
|
70648 |
|
CC1=NC2=C(C=C1)C=C(C=C2)OC |
|
C11H11NO |
|
MFCD00006761 |
|
2-methy-6-methoxyquinolinel, 2-methyl-6-methoxyquinoline, 6-methoxy-2-methyl-quinoline, 6-methoxyquinaldine, acmc-2098xp, pubchem5910, quinaldine, 6-methoxy, quinoline, 6-methoxy-2-methyl, quinoline,6-methoxy-2-methyl |
|
6-methoxy-2-methylquinoline |
Specifications
|
Authentic |
|
Beige to Gray |
|
1g |
|
96.0 |
|
97% |
|
MFCD00006761 |
|
NAGJQQFMJKMXJQ-UHFFFAOYSA-N |
|
6-methoxy-2-methylquinoline |
|
70648 |
|
Fluffy Crystalline Powder |
|
6-Methoxyquinaldine |
|
Glass bottle |
|
61°C to 65°C |
|
1078-28-0 |
|
100.0 |
|
C11H11NO |
|
2-methy-6-methoxyquinolinel, 2-methyl-6-methoxyquinoline, 6-methoxy-2-methyl-quinoline, 6-methoxyquinaldine, acmc-2098xp, pubchem5910, quinaldine, 6-methoxy, quinoline, 6-methoxy-2-methyl, quinoline,6-methoxy-2-methyl |
|
CC1=NC2=C(C=C1)C=C(C=C2)OC |
|
173.21 |
|
173.21 |
|
97% |
Safety and Handling
SDS