Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxypyridine-3-carboxaldehyde, 98%

$109.92 - $354.04

Chemical Identifiers

Chemical Name or Material 6-Methoxypyridine-3-carboxaldehyde
CAS 65873-72-5
Molecular Formula C7H7NO2
Formula Weight 137.14
MDL Number MFCD02683446
Synonym 6-Methoxy-3-nicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2651503

 
alfa aesar™
H2651503
1g Each for $109.92

AAH2651506

 
alfa aesar™
H2651506
5g Each for $354.04
Description & Specifications

Specifications

Formula Weight 137.14
Chemical Name or Material 6-Methoxypyridine-3-carboxaldehyde
Assay 98%
Molecular Formula C7H7NO2
MDL Number MFCD02683446
CAS 65873-72-5
Melting Point 49° to 52°C
Synonym 6-Methoxy-3-nicotinaldehyde
Quantity 1g
Flash Point 107°C (225°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.