missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxypyridine-2-carboxylic Acid, TCI America™

$62.64 - $184.86

Chemical Identifiers

CAS 26893-73-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD06800962
InChI Key KSWBODXXZITTPO-UHFFFAOYSA-N
Synonym 6-methoxypicolinic acid, 6-methoxy-2-pyridinecarboxylic acid, 2-methoxy-6-picolinic acid, 2-methoxy-6-picolinicacid, 2-pyridinecarboxylic acid, 6-methoxy, picolinic acid, 6-methoxy, 6-methoxy-pyridine-2-carboxylic acid, pubchem15165, 6-methoxypyridine-2-carboxylicacid, acmc-1cccg
PubChem CID 570891
IUPAC Name 6-methoxypyridine-2-carboxylic acid
SMILES COC1=CC=CC(=N1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21895G View Documents TCI America
M21895G
5g Each for $184.86
 
M21891G View Documents TCI America
M21891G
1g Each for $62.64
 
Specifications

Chemical Identifiers

26893-73-2
153.137
KSWBODXXZITTPO-UHFFFAOYSA-N
570891
COC1=CC=CC(=N1)C(=O)O
C7H7NO3
MFCD06800962
6-methoxypicolinic acid, 6-methoxy-2-pyridinecarboxylic acid, 2-methoxy-6-picolinic acid, 2-methoxy-6-picolinicacid, 2-pyridinecarboxylic acid, 6-methoxy, picolinic acid, 6-methoxy, 6-methoxy-pyridine-2-carboxylic acid, pubchem15165, 6-methoxypyridine-2-carboxylicacid, acmc-1cccg
6-methoxypyridine-2-carboxylic acid

Specifications

White-Yellow
5g
C7H7NO3
6-methoxypicolinic acid, 6-methoxy-2-pyridinecarboxylic acid, 2-methoxy-6-picolinic acid, 2-methoxy-6-picolinicacid, 2-pyridinecarboxylic acid, 6-methoxy, picolinic acid, 6-methoxy, 6-methoxy-pyridine-2-carboxylic acid, pubchem15165, 6-methoxypyridine-2-carboxylicacid, acmc-1cccg
COC1=CC=CC(=N1)C(=O)O
153.137
153.14
≥98.0% (GC,T)
134°C
26893-73-2
MFCD06800962
KSWBODXXZITTPO-UHFFFAOYSA-N
6-methoxypyridine-2-carboxylic acid
570891
Crystal-Powder at 20°C
6-Methoxypyridine-2-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No