Learn More
6-Methoxypyridine-2-boronic acid pinacol ester, 97%
CAS: 1034297-69-2 | C12H18BNO3 | 235.09 g/mol
$151.82 - $1973.62
Chemical Identifiers
| CAS | 1034297-69-2 |
|---|---|
| Molecular Formula | C12H18BNO3 |
| Molecular Weight (g/mol) | 235.09 |
| MDL Number | MFCD06798266 |
| InChI Key | VYWCRVNLPOVYJH-UHFFFAOYSA-N |
| Synonym | 2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxypyridine-2-boronic acid pinacol ester, pyridine,2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-methoxy pyridine-2-boronic acid pinacol ester, 6-methoxypyridine-2-boronicacidpinacolester, 6-methoxypyridin-2-ylboronic acid pinacol ester, 6-methoxypyridin-2-yl boronic acid pinacol ester, 2-methoxy-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-methoxy-2-pyridinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyloxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine |
| PubChem CID | 17750508 |
| IUPAC Name | 2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2726303
|
Thermo Scientific Chemicals
H2726303 |
1 g |
Each for $151.82
|
|
|||||
|
AAH2726306
|
Thermo Scientific Chemicals
H2726306 |
5 g |
Each for $496.05
|
|
|||||
|
AAH2726314
|
Thermo Scientific Chemicals
H2726314 |
25 g |
Each for $1,973.62
|
|
|||||
Description
It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 1034297-69-2 | |
| 235.09 | |
| VYWCRVNLPOVYJH-UHFFFAOYSA-N | |
| 17750508 | |
| B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)OC |
| C12H18BNO3 | |
| MFCD06798266 | |
| 2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxypyridine-2-boronic acid pinacol ester, pyridine,2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-methoxy pyridine-2-boronic acid pinacol ester, 6-methoxypyridine-2-boronicacidpinacolester, 6-methoxypyridin-2-ylboronic acid pinacol ester, 6-methoxypyridin-2-yl boronic acid pinacol ester, 2-methoxy-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-methoxy-2-pyridinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyloxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine | |
| 2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Specifications
| 1034297-69-2 | |
| C12H18BNO3 | |
| 1 g | |
| Insoluble in water. | |
| B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)OC | |
| 235.09 | |
| 235.09 | |
| 6-Methoxypyridine-2-boronic acid pinacol ester |
| 68°C to 72°C | |
| MFCD06798266 | |
| 2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxypyridine-2-boronic acid pinacol ester, pyridine,2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-methoxy pyridine-2-boronic acid pinacol ester, 6-methoxypyridine-2-boronicacidpinacolester, 6-methoxypyridin-2-ylboronic acid pinacol ester, 6-methoxypyridin-2-yl boronic acid pinacol ester, 2-methoxy-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-methoxy-2-pyridinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyloxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine | |
| VYWCRVNLPOVYJH-UHFFFAOYSA-N | |
| 2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
| 17750508 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only