missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxypyridine-2-boronic acid pinacol ester, 97%

CAS: 1034297-69-2 | C12H18BNO3 | 235.09 g/mol

Supplier:  Thermo Scientific Chemicals H2726306

 View more versions of this product

Catalog No. AAH2726306


Only null left
Explore available promotions

Description

Description

It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate and also in the design, structure-activity relationship, and characterization of the development candidate nvp-hsp990.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

6-Methoxypyridine-2-boronic acid pinacol ester
68°C to 72°C
MFCD06798266
2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxypyridine-2-boronic acid pinacol ester, pyridine,2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-methoxy pyridine-2-boronic acid pinacol ester, 6-methoxypyridine-2-boronicacidpinacolester, 6-methoxypyridin-2-ylboronic acid pinacol ester, 6-methoxypyridin-2-yl boronic acid pinacol ester, 2-methoxy-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-methoxy-2-pyridinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyloxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
VYWCRVNLPOVYJH-UHFFFAOYSA-N
2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
17750508
97%
1034297-69-2
C12H18BNO3
5 g
Insoluble in water.
B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)OC
235.09
235.09

Chemical Identifiers

1034297-69-2
235.09
VYWCRVNLPOVYJH-UHFFFAOYSA-N
17750508
B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)OC
C12H18BNO3
MFCD06798266
2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxypyridine-2-boronic acid pinacol ester, pyridine,2-methoxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-methoxy pyridine-2-boronic acid pinacol ester, 6-methoxypyridine-2-boronicacidpinacolester, 6-methoxypyridin-2-ylboronic acid pinacol ester, 6-methoxypyridin-2-yl boronic acid pinacol ester, 2-methoxy-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-6-methoxy-2-pyridinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methyloxy-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
2-methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only