Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxymethyl-2-thiouracil, 98%

$343.54 - $343.54

Chemical Identifiers

Chemical Name or Material 6-Methoxymethyl-2-thiouracil
CAS 175205-07-9
Molecular Formula C6H8N2O2S
Formula Weight 172.2
MDL Number MFCD00052620
Synonym 4-Hydroxy-6-methoxymethylpyrimidine-2-thiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1855706

  SDS
alfa aesar™
A1855706
5g Each for $343.54

AAA1855703

  SDS
alfa aesar™
A1855703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 6-Methoxymethyl-2-thiouracil
Melting Point 217° to 220°C
Quantity 5g
CAS 175205-07-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C6H8N2O2S
Formula Weight 172.2
MDL Number MFCD00052620
Synonym 4-Hydroxy-6-methoxymethylpyrimidine-2-thiol

Applications
6-Methoxymethyl-2-thiouracil is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.