missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxyindole-2-carboxylic acid, 95%

$82.65 - $396.32

Chemical Identifiers

CAS 16732-73-3
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD01548823
InChI Key XNBGANWAZJWOHS-UHFFFAOYSA-N
Synonym 6-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-methoxy, pubchem1703, zlchem 1017, 6-methoxindole-2-carboxylic acid, indole-2-carboxylic acid, 6-methoxy, asinex-reag bas 06489245, 6-methoxy-1 h-indole-2-carboxylic acid, 6-methoxyindole-2-carboxylic acid, #
PubChem CID 410327
IUPAC Name 6-methoxy-1H-indole-2-carboxylic acid
SMILES COC1=CC2=C(C=C1)C=C(N2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6623903 View Documents Alfa Aesar™
H6623903
1g Each for $82.65
 
AAH6623906 View Documents Alfa Aesar™
H6623906
5g Each for $396.32
 
Specifications

Chemical Identifiers

16732-73-3
191.186
XNBGANWAZJWOHS-UHFFFAOYSA-N
410327
COC1=CC2=C(C=C1)C=C(N2)C(=O)O
C10H9NO3
MFCD01548823
6-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-methoxy, pubchem1703, zlchem 1017, 6-methoxindole-2-carboxylic acid, indole-2-carboxylic acid, 6-methoxy, asinex-reag bas 06489245, 6-methoxy-1 h-indole-2-carboxylic acid, 6-methoxyindole-2-carboxylic acid, #
6-methoxy-1H-indole-2-carboxylic acid

Specifications

198°C to 203°C
16732-73-3
MFCD01548823
6-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-methoxy, pubchem1703, zlchem 1017, 6-methoxindole-2-carboxylic acid, indole-2-carboxylic acid, 6-methoxy, asinex-reag bas 06489245, 6-methoxy-1 h-indole-2-carboxylic acid, 6-methoxyindole-2-carboxylic acid, #
COC1=CC2=C(C=C1)C=C(N2)C(=O)O
191.186
191.19
95%
1g
C10H9NO3
156983
XNBGANWAZJWOHS-UHFFFAOYSA-N
6-methoxy-1H-indole-2-carboxylic acid
410327
Solid
6-Methoxyindole-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning