missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxyflavone 99.0+%, TCI America™

$57.61 - $180.39

Chemical Identifiers

CAS 26964-24-9
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.269
MDL Number MFCD00017322
InChI Key XZQLSABETMKIGG-UHFFFAOYSA-N
Synonym 4h-1-benzopyran-4-one, 6-methoxy-2-phenyl
PubChem CID 147157
IUPAC Name 6-methoxy-2-phenylchromen-4-one
SMILES COC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13461G View Documents Tci America
M13461G
1g Each for $57.61
 
M13465G View Documents Tci America
M13465G
5g Each for $180.39
 
Specifications

Chemical Identifiers

26964-24-9
252.269
XZQLSABETMKIGG-UHFFFAOYSA-N
147157
COC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3
C16H12O3
MFCD00017322
4h-1-benzopyran-4-one, 6-methoxy-2-phenyl
6-methoxy-2-phenylchromen-4-one

Specifications

White-Yellow
1g
C16H12O3
4h-1-benzopyran-4-one, 6-methoxy-2-phenyl
COC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3
252.269
252.27
≥99.0% (GC)
165°C
26964-24-9
MFCD00017322
XZQLSABETMKIGG-UHFFFAOYSA-N
6-methoxy-2-phenylchromen-4-one
147157
Crystal-Powder at 20°C
6-Methoxyflavone
Safety and Handling

Safety and Handling

RTECSNumber : DJ3100438

TSCA : No