missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxyflavone, 98%

$65.96 - $731.14

Chemical Identifiers

CAS 26964-24-9
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.269
MDL Number MFCD00017322
InChI Key XZQLSABETMKIGG-UHFFFAOYSA-N
Synonym 4h-1-benzopyran-4-one, 6-methoxy-2-phenyl
PubChem CID 147157
IUPAC Name 6-methoxy-2-phenylchromen-4-one
SMILES COC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2461603 View Documents Alfa Aesar™
B2461603
1g Each for $65.96
 
AAB2461606 View Documents Alfa Aesar™
B2461606
5g Each for $205.91
 
AAB2461614 View Documents Alfa Aesar™
B2461614
25g Each for $731.14
 
Specifications

Chemical Identifiers

26964-24-9
252.269
XZQLSABETMKIGG-UHFFFAOYSA-N
147157
COC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3
C16H12O3
MFCD00017322
4h-1-benzopyran-4-one, 6-methoxy-2-phenyl
6-methoxy-2-phenylchromen-4-one

Specifications

162°C to 165°C
26964-24-9
MFCD00017322
XZQLSABETMKIGG-UHFFFAOYSA-N
6-methoxy-2-phenylchromen-4-one
147157
98%
1g
C16H12O3
4h-1-benzopyran-4-one, 6-methoxy-2-phenyl
COC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3
252.269
252.27
6-Methoxyflavone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 248-144-5

RTECSNumber : DJ3100438

TSCA : No

Recommended Storage : Ambient temperatures