missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxyflavanone, 98%

$69.96 - $891.68

Chemical Identifiers

CAS 3034-04-6
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.29
MDL Number MFCD00017484
InChI Key YURQMHCZHLMHIB-UHFFFAOYNA-N
Synonym 2,3-dihydro-6-methoxy-2-phenyl-4h-1-benzopyran-4-one
PubChem CID 97860
IUPAC Name 6-methoxy-2-phenyl-2,3-dihydrochromen-4-one
SMILES COC1=CC2=C(C=C1)OC(CC2=O)C3=CC=CC=C3
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2524003 View Documents Alfa Aesar™
B2524003
1g Each for $69.96
 
AAB2524006 View Documents Alfa Aesar™
B2524006
5g Each for $216.38
 
AAB2524014 View Documents Alfa Aesar™
B2524014
25g Each for $891.68
 
Specifications

Chemical Identifiers

3034-04-6
254.29
YURQMHCZHLMHIB-UHFFFAOYNA-N
97860
COC1=CC2=C(C=C1)OC(CC2=O)C3=CC=CC=C3
C16H14O3
MFCD00017484
2,3-dihydro-6-methoxy-2-phenyl-4h-1-benzopyran-4-one
6-methoxy-2-phenyl-2,3-dihydrochromen-4-one

Specifications

141°C to 143°C
3034-04-6
MFCD00017484
1-2-trifluoromethyl phenyl methanamine
C1=CC=C(C(=C1)CN)C(F)(F)F
175.15
254.29
6-Methoxyflavanone
1g
C8H8F3N
248912
ZSKQIFWUTUZAGF-UHFFFAOYSA-N
[2-(trifluoromethyl)phenyl]methanamine
76447
98%
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures