missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxy-3-pyridinecarboxaldehyde, TCI America™

$99.31 - $334.59

Chemical Identifiers

CAS 65873-72-5
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD02683446
InChI Key CTAIEPPAOULMFY-UHFFFAOYSA-N
Synonym 6-methoxynicotinaldehyde, 6-methoxy-3-pyridinecarboxaldehyde, 5-formyl-2-methoxypyridine, 2-methoxypyridine-5-carboxaldehyde, 5-formyl-2-methoxy-pyridine, 6-methoxy-pyridine-3-carbaldehyde, 6-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-methoxy, 6-methoxy-3-nicotinaldehyde, pubchem5152
PubChem CID 3364576
IUPAC Name 6-methoxypyridine-3-carbaldehyde
SMILES COC1=NC=C(C=C1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23475G View Documents TCI America
M23475G
5g Each for $334.59
 
M23471G View Documents TCI America
M23471G
1g Each for $99.31
 
Specifications

Chemical Identifiers

65873-72-5
137.138
CTAIEPPAOULMFY-UHFFFAOYSA-N
3364576
COC1=NC=C(C=C1)C=O
C7H7NO2
MFCD02683446
6-methoxynicotinaldehyde, 6-methoxy-3-pyridinecarboxaldehyde, 5-formyl-2-methoxypyridine, 2-methoxypyridine-5-carboxaldehyde, 5-formyl-2-methoxy-pyridine, 6-methoxy-pyridine-3-carbaldehyde, 6-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-methoxy, 6-methoxy-3-nicotinaldehyde, pubchem5152
6-methoxypyridine-3-carbaldehyde

Specifications

White
5g
C7H7NO2
6-methoxynicotinaldehyde, 6-methoxy-3-pyridinecarboxaldehyde, 5-formyl-2-methoxypyridine, 2-methoxypyridine-5-carboxaldehyde, 5-formyl-2-methoxy-pyridine, 6-methoxy-pyridine-3-carbaldehyde, 6-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-methoxy, 6-methoxy-3-nicotinaldehyde, pubchem5152
COC1=NC=C(C=C1)C=O
137.138
137.14
≥98.0% (GC)
51°C
65873-72-5
MFCD02683446
CTAIEPPAOULMFY-UHFFFAOYSA-N
6-methoxypyridine-3-carbaldehyde
3364576
Crystal-Powder at 20°C
6-Methoxy-3-pyridinecarboxaldehyde
Safety and Handling

Safety and Handling

TSCA : No