Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxy-2-tetralone, tech. 90%

$63.28 - $170.27

Chemical Identifiers

Chemical Name or Material 6-Methoxy-2-tetralone
CAS 2472-22-2
Purity Grade Technical Grade
Molecular Formula C11H12O2
Formula Weight 176.22
MDL Number MFCD00001729
Beilstein 513418
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1100703

  SDS
alfa aesar™
L1100703
1g Each for $170.27

AAL11007MD

  SDS
alfa aesar™
L11007MD
250mg Each for $63.28
Description & Specifications

Specifications

Formula Weight 176.22
Assay 90%
Flash Point >110°C (230°F)
Chemical Name or Material 6-Methoxy-2-tetralone
MDL Number MFCD00001729
Beilstein 513418
Boiling Point 114° to 116°C (0.2mmHg)
CAS 2472-22-2
Molecular Formula C11H12O2
Purity Grade Technical Grade
Quantity 1g
Recommended Storage 2° to 8°C
Melting Point ≈35°C
Safety and Handling

2° to 8°C