missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxy-2-tetralone, tech. 90%

Manufacturer:  Alfa Aesar™ L1100703

 View more versions of this product

Catalog No. AAL1100703



Specifications

Specifications

6-Methoxy-2-tetralone
114°C to 116°C (0.2mmHg)
1g
C11H12O2
513418
RMRKDYNVZWKAFP-UHFFFAOYSA-N
6-methoxy-3,4-dihydro-1H-naphthalen-2-one
75582
90%
>110°C (230°F)
~35°C
2472-22-2
MFCD00001729
6-methoxy-2-tetralone, 6-methoxy-3,4-dihydronaphthalen-2 1h-one, 6-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 6-methoxyl-2-tetralone, 3,4-dihydro-6-methoxy-2 1h-naphthalenone, 6-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-6-methoxynaphthalen-2 1h-one, 6-methoxy-1,3,4-trihydronaphthalen-2-one
COC1=CC2=C(CC(=O)CC2)C=C1
176.215
176.22
Technical

Chemical Identifiers

2472-22-2
176.215
RMRKDYNVZWKAFP-UHFFFAOYSA-N
75582
COC1=CC2=C(CC(=O)CC2)C=C1
C11H12O2
MFCD00001729
6-methoxy-2-tetralone, 6-methoxy-3,4-dihydronaphthalen-2 1h-one, 6-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 6-methoxyl-2-tetralone, 3,4-dihydro-6-methoxy-2 1h-naphthalenone, 6-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-6-methoxynaphthalen-2 1h-one, 6-methoxy-1,3,4-trihydronaphthalen-2-one
6-methoxy-3,4-dihydro-1H-naphthalen-2-one
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents