missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxy-2-tetralone, TCI America™

$166.53 - $627.92

Chemical Identifiers

CAS 2472-22-2
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00001729
InChI Key RMRKDYNVZWKAFP-UHFFFAOYSA-N
Synonym 6-methoxy-2-tetralone, 6-methoxy-3,4-dihydronaphthalen-2 1h-one, 6-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 6-methoxyl-2-tetralone, 3,4-dihydro-6-methoxy-2 1h-naphthalenone, 6-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-6-methoxynaphthalen-2 1h-one, 6-methoxy-1,3,4-trihydronaphthalen-2-one
PubChem CID 75582
IUPAC Name 6-methoxy-3,4-dihydro-1H-naphthalen-2-one
SMILES COC1=CC2=C(CC(=O)CC2)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13175G View Documents TCI America
M13175G
5g Each for $627.92
 
M13171G View Documents TCI America
M13171G
1g Each for $166.53
 
Specifications

Chemical Identifiers

2472-22-2
176.215
RMRKDYNVZWKAFP-UHFFFAOYSA-N
75582
COC1=CC2=C(CC(=O)CC2)C=C1
C11H12O2
MFCD00001729
6-methoxy-2-tetralone, 6-methoxy-3,4-dihydronaphthalen-2 1h-one, 6-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 6-methoxyl-2-tetralone, 3,4-dihydro-6-methoxy-2 1h-naphthalenone, 6-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-6-methoxynaphthalen-2 1h-one, 6-methoxy-1,3,4-trihydronaphthalen-2-one
6-methoxy-3,4-dihydro-1H-naphthalen-2-one

Specifications

Undesignated
2472-22-2
MFCD00001729
RMRKDYNVZWKAFP-UHFFFAOYSA-N
6-methoxy-3,4-dihydro-1H-naphthalen-2-one
75582
Crystal at 20°C
6-Methoxy-2-tetralone
5g
C11H12O2
6-methoxy-2-tetralone, 6-methoxy-3,4-dihydronaphthalen-2 1h-one, 6-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 6-methoxyl-2-tetralone, 3,4-dihydro-6-methoxy-2 1h-naphthalenone, 6-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-6-methoxynaphthalen-2 1h-one, 6-methoxy-1,3,4-trihydronaphthalen-2-one
COC1=CC2=C(CC(=O)CC2)C=C1
176.215
176.22
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator