Please login to your online account to display your discounted pricing

6-Methoxy-2-tetralone, 90%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC298922500

 View more versions of this product

Catalog No. AC298922500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥88%
Physical Form Crystalline Low Melting Solid
Formula Weight 176.22
Quantity 250mg
Color Light yellow-beige to orange
Melting Point 28° to 35°C
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C11H12O2
Infrared Spectrum Authentic
MDL Number MFCD00001729
Packaging Glass bottle
Chemical Name or Material 6-Methoxy-2-tetralone
CAS 2472-22-2
Synonym 3, 4-Dihydro-6-methoxy-2(1H)-naphthalenone

Specifications

Chemical Name or Material 6-Methoxy-2-tetralone
CAS 2472-22-2
CAS Min % 88.0
CAS Max % 100.0
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C11H12O2
Formula Weight 176.22
MDL Number MFCD00001729
Synonym 3, 4-Dihydro-6-methoxy-2(1H)-naphthalenone