missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxy-2-tetralone, 90%, ACROS Organics™

Chemical Identifiers

CAS 2472-22-2
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C11H12O2
Formula Weight 176.22
MDL Number MFCD00001729
Synonym 3, 4-Dihydro-6-methoxy-2(1H)-naphthalenone
Chemical Name or Material 6-Methoxy-2-tetralone
Assay ≥88%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC298922500 SDS Thermo Fisher Scientific
AC298922500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC298920050 SDS Thermo Fisher Scientific
AC298920050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC298920010 SDS Fisher Scientific Acros
298920010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

2472-22-2
100.0
C11H12O2
MFCD00001729
6-Methoxy-2-tetralone
88.0
90%
176.22
3, 4-Dihydro-6-methoxy-2(1H)-naphthalenone
≥88%

Specifications

Crystalline Low Melting Solid
250mg
28° to 35°C
100.0
90%
Authentic
3, 4-Dihydro-6-methoxy-2(1H)-naphthalenone
6-Methoxy-2-tetralone
176.22
Light yellow-beige to orange
88.0
C11H12O2
MFCD00001729
Glass bottle
2472-22-2
≥88%