Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxy-2-naphthylacetic acid, 99%

Chemical Identifiers

Chemical Name or Material 6-Methoxy-2-naphthylacetic acid
CAS 23981-47-7
Formula Weight 216.24
MDL Number MFCD00033105
Synonym 6-MNA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67138LB0

 
alfa aesar™
J67138LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67138MA

 
alfa aesar™
J67138MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67138MF

 
alfa aesar™
J67138MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.24
Melting Point 171° to 173°C
Chemical Name or Material 6-Methoxy-2-naphthylacetic acid
Physical Form Crystalline Solid
Assay 99%
MDL Number MFCD00033105
CAS 23981-47-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number QJ1045000
Quantity 5mg
Synonym 6-MNA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.