Learn More
6-Methoxy-2-naphthaldehyde, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2394314
Description
6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Methoxy-2-naphthaldehyde is used as a diagnostic reagent in tumor studies involving aldehyde dehydrogenase enzymes. In addition it is used in organic synthesis reactions forming fluorescent substrates for inhibition studies relating to hypertension and vascular inflammation. It is also used as intermediate of Nabumetone.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to air. Incompatible with oxidizing agents.
Specifications
6-Methoxy-2-naphthaldehyde | |
80°C to 82°C | |
MFCD00081152 | |
1945989 | |
6-methoxy-2-naphthaldehyde, 6-methoxy-2-naphthalaldehyde, unii-9244l5l5zy, 6-methoxy-naphthalene-2-carbaldehyde, 2-naphthalenecarboxaldehyde, 6-methoxy, pubchem13704, acmc-1ah4r, 6-methoxy-2-napthaldehyde, dsstox_cid_31510, dsstox_rid_97395 | |
COC1=CC=C2C=C(C=O)C=CC2=C1 | |
186.21 | |
186.21 |
3453-33-6 | |
C12H10O2 | |
25 g | |
Air Sensitive | |
VZBLASFLFFMMCM-UHFFFAOYSA-N | |
6-methoxynaphthalene-2-carbaldehyde | |
76991 | |
99% |
Chemical Identifiers
3453-33-6 | |
186.21 | |
VZBLASFLFFMMCM-UHFFFAOYSA-N | |
76991 | |
COC1=CC=C2C=C(C=O)C=CC2=C1 |
C12H10O2 | |
MFCD00081152 | |
6-methoxy-2-naphthaldehyde, 6-methoxy-2-naphthalaldehyde, unii-9244l5l5zy, 6-methoxy-naphthalene-2-carbaldehyde, 2-naphthalenecarboxaldehyde, 6-methoxy, pubchem13704, acmc-1ah4r, 6-methoxy-2-napthaldehyde, dsstox_cid_31510, dsstox_rid_97395 | |
6-methoxynaphthalene-2-carbaldehyde |
Safety and Handling
EINECSNumber : 222-377-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only