missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H54712MD

 View more versions of this product

Catalog No. AAH54712MD



Specifications

Specifications

6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester
1080028-73-4
MFCD12923398
CWJKZNNXSPVPAY-UHFFFAOYSA-N
6-methoxy-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
67487482
96%
250mg
C13H20BNO3
6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxy-2-methylpyridine-3-boronic acid pinacol ester, \rtf1\ansi\deff0 \fonttbl \f0\fnil\fcharset0 ms sans serif;, 6-methoxy-2-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxy-2-methyl-3-boronic acid pinacol ester pyridine, 6-methoxy-2-methyl-pyridine-3-boronic acid pinacol ester, 2-methyl-6-methoxypyridine-3-boronic acid pinacol ester, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(C=C2)OC)C
249.117
249.11

Chemical Identifiers

1080028-73-4
249.117
CWJKZNNXSPVPAY-UHFFFAOYSA-N
67487482
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(C=C2)OC)C
C13H20BNO3
MFCD12923398
6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxy-2-methylpyridine-3-boronic acid pinacol ester, \rtf1\ansi\deff0 \fonttbl \f0\fnil\fcharset0 ms sans serif;, 6-methoxy-2-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-methoxy-2-methyl-3-boronic acid pinacol ester pyridine, 6-methoxy-2-methyl-pyridine-3-boronic acid pinacol ester, 2-methyl-6-methoxypyridine-3-boronic acid pinacol ester, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 6-methoxy-2-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
6-methoxy-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents