missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5471203

 View more versions of this product

Catalog No. AAH5471203



Specifications

Specifications

6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester
1080028-73-4
MFCD12923398
CWJKZNNXSPVPAY-UHFFFAOYSA-N
6-methoxy-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
67487482
96%
1g
C13H20BNO3
2-methyl-6-methoxypyridine-3-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(C=C2)OC)C
249.117
249.11

Chemical Identifiers

1080028-73-4
249.117
CWJKZNNXSPVPAY-UHFFFAOYSA-N
67487482
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(C=C2)OC)C
C13H20BNO3
MFCD12923398
2-methyl-6-methoxypyridine-3-boronic acid pinacol ester
6-methoxy-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents