missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene 98.0+%, TCI America™
$136.03 - $471.16
Chemical Identifiers
CAS | 63675-74-1 |
---|---|
Molecular Formula | C16H14O2S |
Molecular Weight (g/mol) | 270.35 |
MDL Number | MFCD02083138 |
InChI Key | HRWAGCVMOGWQJF-UHFFFAOYSA-N |
PubChem CID | 850941 |
IUPAC Name | 6-methoxy-2-(4-methoxyphenyl)-1-benzothiophene |
SMILES | COC1=CC=C(C=C1)C1=CC2=CC=C(OC)C=C2S1 |
Chemical Identifiers
63675-74-1 | |
270.35 | |
HRWAGCVMOGWQJF-UHFFFAOYSA-N | |
6-methoxy-2-(4-methoxyphenyl)-1-benzothiophene |
C16H14O2S | |
MFCD02083138 | |
850941 | |
COC1=CC=C(C=C1)C1=CC2=CC=C(OC)C=C2S1 |
Specifications
63675-74-1 | |
C16H14O2S | |
5 g | |
COC1=CC=C(C=C1)C1=CC2=CC=C(OC)C=C2S1 | |
270.35 | |
270.35 | |
Crystalline Powder |
198°C | |
MFCD02083138 | |
HRWAGCVMOGWQJF-UHFFFAOYSA-N | |
6-methoxy-2-(4-methoxyphenyl)-1-benzothiophene | |
850941 | |
≥98.0% (GC) | |
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene |
Safety and Handling
TSCA : No