missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene 98.0+%, TCI America™
Supplier: TCI America M234525G
Specifications
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene | |
198°C | |
MFCD02083138 | |
HRWAGCVMOGWQJF-UHFFFAOYSA-N | |
6-methoxy-2-(4-methoxyphenyl)-1-benzothiophene | |
850941 | |
≥98.0% (GC) |
63675-74-1 | |
C16H14O2S | |
25 g | |
COC1=CC=C(C=C1)C1=CC2=CC=C(OC)C=C2S1 | |
270.35 | |
270.35 | |
Crystalline Powder |
Chemical Identifiers
63675-74-1 | |
270.35 | |
HRWAGCVMOGWQJF-UHFFFAOYSA-N | |
6-methoxy-2-(4-methoxyphenyl)-1-benzothiophene |
C16H14O2S | |
MFCD02083138 | |
850941 | |
COC1=CC=C(C=C1)C1=CC2=CC=C(OC)C=C2S1 |
Safety and Handling
TSCA : No