Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxy-2,4-bis(trifluoromethyl)benzoic acid, 97%

$145.31 - $431.00

Chemical Identifiers

Chemical Name or Material 6-Methoxy-2,4-bis(trifluoromethyl)benzoic acid
CAS 180134-15-0
Molecular Formula C10H6F6O3
Formula Weight 288.14
MDL Number MFCD00728738
Synonym 2,4-Bis(trifluoromethyl)-6-methoxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2644803

  SDS
alfa aesar™
H2644803
1g Each for $145.31

AAH2644806

  SDS
alfa aesar™
H2644806
5g Each for $431.00
Description & Specifications

Specifications

Formula Weight 288.14
Chemical Name or Material 6-Methoxy-2,4-bis(trifluoromethyl)benzoic acid
Assay 97%
Molecular Formula C10H6F6O3
MDL Number MFCD00728738
CAS 180134-15-0
Synonym 2,4-Bis(trifluoromethyl)-6-methoxybenzoic acid
Melting Point 156° to 159°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.