Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxy-2(3H)-benzoxazolone, 98+%

$69.58 - $190.86

Chemical Identifiers

Chemical Name or Material 6-Methoxy-2(3H)-benzoxazolone
CAS 532-91-2
Molecular Formula C8H7NO3
Formula Weight 165.15
MDL Number MFCD00051726
Beilstein 141041
Synonym 6-Methoxy-2-benzoxazolinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0534903

 
alfa aesar™
L0534903
1g Each for $190.86

AAL05349MD

 
alfa aesar™
L05349MD
250mg Each for $69.58
Description & Specifications

Specifications

Formula Weight 165.15
Chemical Name or Material 6-Methoxy-2(3H)-benzoxazolone
Assay ≥98%
Molecular Formula C8H7NO3
CAS 532-91-2
MDL Number MFCD00051726
Beilstein 141041
Synonym 6-Methoxy-2-benzoxazolinone
RTECS Number DM5275200
Melting Point 152° to 156°C
Quantity 1g