missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxy-1-tetralone, TCI America™

Manufacturer:  TCI America M078725G

 View more versions of this product

Catalog No. M078725G



Specifications

Specifications

6-Methoxy-1-tetralone
79°C
1078-19-9
MFCD00001695
MNALUTYMBUBKNX-UHFFFAOYSA-N
6-methoxy-3,4-dihydro-2H-naphthalen-1-one
14112
Crystal-Powder at 20°C
Undesignated
25g
C11H12O2
6-methoxy-1-tetralone, 6-methoxy-3,4-dihydronaphthalen-1 2h-one, 6-methoxytetralone, 6-methoxy-alpha-tetralone, 1 2h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-methoxy-3,4-dihydro-1 2h-naphthalenone, 6-methoxytetralin-1-one, 3,4-dihydro-6-methoxy-1 2h-naphthalenone, 6-methoxy-.alpha.-tetralone
COC1=CC2=C(C=C1)C(=O)CCC2
176.215
176.22
≥99.0% (GC)

Chemical Identifiers

1078-19-9
176.215
MNALUTYMBUBKNX-UHFFFAOYSA-N
14112
COC1=CC2=C(C=C1)C(=O)CCC2
C11H12O2
MFCD00001695
6-methoxy-1-tetralone, 6-methoxy-3,4-dihydronaphthalen-1 2h-one, 6-methoxytetralone, 6-methoxy-alpha-tetralone, 1 2h-naphthalenone, 3,4-dihydro-6-methoxy, 6-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 6-methoxy-3,4-dihydro-1 2h-naphthalenone, 6-methoxytetralin-1-one, 3,4-dihydro-6-methoxy-1 2h-naphthalenone, 6-methoxy-.alpha.-tetralone
6-methoxy-3,4-dihydro-2H-naphthalen-1-one
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0371

TSCA : Yes

Documents