Please login to your online account to display your discounted pricing

6-Methoxy-1-tetralone, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC126120250

 View more versions of this product

Catalog No. AC126120250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 176.21
Quantity 25g
Color Yellow to light brown
Beilstein 09, II, 889
Merck Index 15, 6074
Melting Point 77° to 79°C
CAS Max % 100.0
Chemical Name or Material 6-Methoxy-1-tetralone
Assay Percent Range 99%
Molecular Formula C11H12O2
CAS 1078-19-9
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00001695
Synonym 3, 4-Dihydro-6-methoxy-1(2H)-naphthalenone
CAS Min % 98.5

Specifications

Chemical Name or Material 6-Methoxy-1-tetralone
CAS 1078-19-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H12O2
Formula Weight 176.21
MDL Number MFCD00001695
Synonym 3, 4-Dihydro-6-methoxy-1(2H)-naphthalenone