missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Methoxy-1,2,3,4-tetrahydronaphthalene 98.0+%, TCI America™
Supplier: TCI America M076525ML
Specifications
6-Methoxy-1,2,3,4-tetrahydronaphthalene | |
Yellow | |
C11H14O | |
25 mL | |
SUFDFKRJYKNTFH-UHFFFAOYSA-N | |
6-methoxy-1,2,3,4-tetrahydronaphthalene | |
74414 | |
≥98.0% (GC) |
1730-48-9 | |
131°C | |
MFCD00001737 | |
6-methoxytetralin, naphthalene, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxynaphthalene, unii-d0xg2umz29, d0xg2umz29, methyl 1,2,3,4-tetrahydro-6-naphthyl ether, acmc-20aj41, 1,3,4-tetrahydro-6-methoxynaphthalene, 6-methoxy-1,3,4-tetrahydronaphthalene, 7-methoxy-3,4-dihydro-1h-naphthalene | |
COC1=CC=C2CCCCC2=C1 | |
162.23 | |
162.23 | |
Liquid |
Chemical Identifiers
1730-48-9 | |
162.23 | |
SUFDFKRJYKNTFH-UHFFFAOYSA-N | |
74414 | |
COC1=CC=C2CCCCC2=C1 |
C11H14O | |
MFCD00001737 | |
6-methoxytetralin, naphthalene, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxynaphthalene, unii-d0xg2umz29, d0xg2umz29, methyl 1,2,3,4-tetrahydro-6-naphthyl ether, acmc-20aj41, 1,3,4-tetrahydro-6-methoxynaphthalene, 6-methoxy-1,3,4-tetrahydronaphthalene, 7-methoxy-3,4-dihydro-1h-naphthalene | |
6-methoxy-1,2,3,4-tetrahydronaphthalene |
Safety and Handling
EINECSNumber : (4)-1394
TSCA : Yes