missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Methoxy-1,2,3,4-tetrahydroisoquinoline, TCI America™

$207.78 - $623.34

Chemical Identifiers

CAS 42923-77-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD01717129
InChI Key YYTAYINRPUJPNH-UHFFFAOYSA-N
Synonym Longimammatine
PubChem CID 39356
IUPAC Name 6-methoxy-1,2,3,4-tetrahydroisoquinoline
SMILES COC1=CC2=C(CNCC2)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27815G View Documents TCI America
M27815G
5g Each for $623.34
 
M27811G View Documents TCI America
M27811G
1g Each for $207.78
 
Specifications

Chemical Identifiers

42923-77-3
163.22
YYTAYINRPUJPNH-UHFFFAOYSA-N
39356
COC1=CC2=C(CNCC2)C=C1
C10H13NO
MFCD01717129
Longimammatine
6-methoxy-1,2,3,4-tetrahydroisoquinoline

Specifications

144°C
5g
C10H13NO
Longimammatine
COC1=CC2=C(CNCC2)C=C1
163.22
163.22
≥96.0% (GC,T)
Yellow
42923-77-3
MFCD01717129
YYTAYINRPUJPNH-UHFFFAOYSA-N
6-methoxy-1,2,3,4-tetrahydroisoquinoline
39356
Clear Liquid at 20°C
6-Methoxy-1,2,3,4-tetrahydroisoquinoline
Safety and Handling

Safety and Handling

RTECSNumber : NX5082000

TSCA : No