Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 95%

$113.76 - $317.93

Chemical Identifiers

Chemical Name or Material 6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
CAS 57196-62-0
Molecular Formula C10H13NO·HCl
Formula Weight 199.68
MDL Number MFCD01717328
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3191003

  SDS
alfa aesar™
H3191003
1g Each for $317.93

AAH31910MD

  SDS
alfa aesar™
H31910MD
250mg Each for $113.76
Description & Specifications

Specifications

Formula Weight 199.68
Assay 95%
Molecular Formula C10H13NO·HCl
MDL Number MFCD01717328
Melting Point 233° to 236°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material 6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
CAS 57196-62-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.