Please login to your online account to display your discounted pricing

6-Mercaptopurine-9-D-riboside, 99+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  226530010

 View more versions of this product

Catalog No. AC226530010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Very Fine Crystalline Powder
Formula Weight 284.28
Color White
Quantity 1g
EINECS Number 209-371-5
Specific Rotation Condition −73° (25°C c=1,0.1N NaOH)
Melting Point 218° to 221°C
RTECS Number UP0710000
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 6-Mercaptopurine-9-β-D-ribofuranoside
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 6-Mercaptopurine-9-D-riboside
Name Note 99+%
Specific Rotation -72° to -74° (25°C, 589nm) (c=1, 0.1N NaOH)
Assay Percent Range 99+%
Molecular Formula C10H12N4O4S
MDL Number MFCD00005743
CAS 574-25-4
Health Hazard 2 GHS H Statement:
Harmful if swallowed.

Specifications

Chemical Name or Material 6-Mercaptopurine-9-D-riboside
Name Note 99+%
CAS 574-25-4
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H12N4O4S
Formula Weight 284.28
MDL Number MFCD00005743
Synonym 6-Mercaptopurine-9-β-D-ribofuranoside
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents