Please login to your online account to display your discounted pricing

6-Mercaptopurine-9-D-riboside, 99+%, ACROS Organics™

$21.96 - $21.96

Chemical Identifiers

Chemical Name or Material 6-Mercaptopurine-9-D-riboside
Name Note 99+%
CAS 574-25-4
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H12N4O4S
Formula Weight 284.28
MDL Number MFCD00005743
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC226531000

  SDS
thermo fisher scientific
226531000
Glass bottle 100mg Each for $21.96
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Very Fine Crystalline Powder
Color White
Formula Weight 284.28
Quantity 100mg
RTECS Number UP0710000
EINECS Number 209-371-5
Melting Point 218° to 221°C
Specific Rotation Condition −73° (25°C c=1,0.1N NaOH)
CAS Max % 100.0
CAS Min % 99.0
Synonym 6-Mercaptopurine-9-β-D-ribofuranoside
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Specific Rotation -72° to -74° (25°C, 589nm) (c=1, 0.1N NaOH)
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Molecular Formula C10H12N4O4S
CAS 574-25-4
Chemical Name or Material 6-Mercaptopurine-9-D-riboside
Name Note 99+%
Assay Percent Range 99+%
MDL Number MFCD00005743