Learn More
6-Maleimidohexanoic acid, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 4638406
Description
6-Maleimidohexanoic acid is used as a probe for introducing maleimides groups into biomolecules and active pharmaceutical ingredients. Further, it is used with N-hydroxysuccinimide ester as a bifunctional cross-linking reagent. In addition to this, it acts as a probe for thiol groups in membrane proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
6-Maleimidohexanoic acid | |
White to Yellow | |
MFCD00043140 | |
Moisture sensitive | |
Soluble in water,methanol,ethanol,dimethyl sulfoxide. Insoluble in ether. | |
C1=CC(=O)N(C1=O)CCCCCC(=O)O | |
211.217 | |
211.22 |
55750-53-3 | |
C10H13NO4 | |
5 g | |
6-maleimidocaproic acid, 6-maleimidohexanoic acid, 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoic acid, 6-maleimidocaproicacid, 1h-pyrrole-1-hexanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 5-cooh, n-maleoyl-6-aminocaproic acid, n-maleoyl-6-aminohexanoic acid, 1h-pyrrole-1-hexanoicacid, 2,5-dihydro-2,5-dioxo, 6-2,5-dioxopyrrol-1-yl hexanoic acid | |
WOJKKJKETHYEAC-UHFFFAOYSA-N | |
6-(2,5-dioxopyrrol-1-yl)hexanoic acid | |
573683 | |
Powder |
Chemical Identifiers
55750-53-3 | |
211.217 | |
WOJKKJKETHYEAC-UHFFFAOYSA-N | |
573683 | |
C1=CC(=O)N(C1=O)CCCCCC(=O)O |
C10H13NO4 | |
MFCD00043140 | |
6-maleimidocaproic acid, 6-maleimidohexanoic acid, 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoic acid, 6-maleimidocaproicacid, 1h-pyrrole-1-hexanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 5-cooh, n-maleoyl-6-aminocaproic acid, n-maleoyl-6-aminohexanoic acid, 1h-pyrrole-1-hexanoicacid, 2,5-dihydro-2,5-dioxo, 6-2,5-dioxopyrrol-1-yl hexanoic acid | |
6-(2,5-dioxopyrrol-1-yl)hexanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only