missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Maleimidohexanoic acid

Manufacturer:  Alfa Aesar™ 4638422

 View more versions of this product

Catalog No. AA4638422



Specifications

Specifications

6-Maleimidohexanoic acid
White to Yellow
55750-53-3
MFCD00043140
WOJKKJKETHYEAC-UHFFFAOYSA-N
6-(2,5-dioxopyrrol-1-yl)hexanoic acid
573683
Powder
Moisture Sensitive
100g
C10H13NO4
6-Maleimidocaproic acid; MCA
C1=CC(=O)N(C1=O)CCCCCC(=O)O
211.217
211.22

Chemical Identifiers

55750-53-3
211.217
WOJKKJKETHYEAC-UHFFFAOYSA-N
573683
C1=CC(=O)N(C1=O)CCCCCC(=O)O
C10H13NO4
MFCD00043140
6-Maleimidocaproic acid; MCA
6-(2,5-dioxopyrrol-1-yl)hexanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2°C to 8°C

SDS Documents