missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Maleimidohexanehydrazide Trifluoroacetate, TCI America™

$239.86 - $718.06

Chemical Identifiers

CAS 151038-94-7
Molecular Formula C12H16F3N3O5
Molecular Weight (g/mol) 339.271
MDL Number MFCD07357647
InChI Key DRURMGRBAVYWCL-UHFFFAOYSA-N
Synonym epsilon-Maleimidocaproyl Hydrazide Trifluoroacetate, EMCH
PubChem CID 23509306
IUPAC Name 6-(2,5-dioxopyrrol-1-yl)hexanehydrazide;2,2,2-trifluoroacetic acid
SMILES C1=CC(=O)N(C1=O)CCCCCC(=O)NN.C(=O)(C(F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M273550MG View Documents TCI America
M273550MG
50mg Each for $239.86
 
M2735250MG View Documents TCI America
M2735250MG
250mg Each for $718.06
 
Specifications

Chemical Identifiers

151038-94-7
339.271
DRURMGRBAVYWCL-UHFFFAOYSA-N
23509306
C1=CC(=O)N(C1=O)CCCCCC(=O)NN.C(=O)(C(F)(F)F)O
C12H16F3N3O5
MFCD07357647
epsilon-Maleimidocaproyl Hydrazide Trifluoroacetate, EMCH
6-(2,5-dioxopyrrol-1-yl)hexanehydrazide;2,2,2-trifluoroacetic acid

Specifications

White-Yellow
151038-94-7
MFCD07357647
DRURMGRBAVYWCL-UHFFFAOYSA-N
6-(2,5-dioxopyrrol-1-yl)hexanehydrazide;2,2,2-trifluoroacetic acid
23509306
Crystal-Powder at 20°C
6-Maleimidohexanehydrazide Trifluoroacetate
50mg
C12H16F3N3O5
epsilon-Maleimidocaproyl Hydrazide Trifluoroacetate, EMCH
C1=CC(=O)N(C1=O)CCCCCC(=O)NN.C(=O)(C(F)(F)F)O
339.271
339.27
≥95.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)