missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Maleimidocaproic acid N-succinimidyl ester, 95%

Manufacturer:  Alfa Aesar™ H65996MD

 View more versions of this product

Catalog No. AAH65996MD



Specifications

Specifications

6-Maleimidocaproic acid N-succinimidyl ester
White to Brown
250mg
C14H16N2O6
1499815
VLARLSIGSPVYHX-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 6-(2,5-dioxopyrrol-1-yl)hexanoate
5091655
Solid
Light and moisture sensitive; Hygroscopic
69°C to 74°C
55750-63-5
MFCD00043043
1h-pyrrole-2,5-dione, 1-6-2,5-dioxo-1-pyrrolidinyl oxy-6-oxohexyl, 2,5-dioxopyrrolidin-1-yl 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoate, 2,5-dioxopyrrolidin-1-yl 6-2,5-dioxopyrrol-1-yl hexanoate, 6-maleimidohexanoic acid n-hydroxysuccinimide ester, emcs, n-6-maleimidocaproyloxy succinimide, n-succinimidyl 6-maleimidocaproate, n-succinimidyl 6-maleimidohexanoate, nmicos
C1CC(=O)N(C1=O)OC(=O)CCCCCN2C(=O)C=CC2=O
308.29
308.29
95%

Chemical Identifiers

55750-63-5
308.29
VLARLSIGSPVYHX-UHFFFAOYSA-N
5091655
C1CC(=O)N(C1=O)OC(=O)CCCCCN2C(=O)C=CC2=O
C14H16N2O6
MFCD00043043
1h-pyrrole-2,5-dione, 1-6-2,5-dioxo-1-pyrrolidinyl oxy-6-oxohexyl, 2,5-dioxopyrrolidin-1-yl 6-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl hexanoate, 2,5-dioxopyrrolidin-1-yl 6-2,5-dioxopyrrol-1-yl hexanoate, 6-maleimidohexanoic acid n-hydroxysuccinimide ester, emcs, n-6-maleimidocaproyloxy succinimide, n-succinimidyl 6-maleimidocaproate, n-succinimidyl 6-maleimidohexanoate, nmicos
(2,5-dioxopyrrolidin-1-yl) 6-(2,5-dioxopyrrol-1-yl)hexanoate
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Nitrogen

Documents