Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Ketoprostaglandin F1alpha, 98%

Chemical Identifiers

Chemical Name or Material 6-Ketoprostaglandin F1alpha
CAS 58962-34-8
Formula Weight 370.49
MDL Number MFCD00135222
Synonym 6-Keto-PGF1α
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67057MCR

  SDS
alfa aesar™
J67057MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 370.49
Chemical Name or Material 6-Ketoprostaglandin F1alpha
Assay 98%
Physical Form Solid
MDL Number MFCD00135222
CAS 58962-34-8
Synonym 6-Keto-PGF1α
Health Hazard 1 Warning
Color White to off-white
Quantity 1mg
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
RTECS Number UK8428300
Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.

-30° to -10°C