Please login to your online account to display your discounted pricing

Alfa Aesar™ 6-Iodouridine, 96%

Chemical Identifiers

Chemical Name or Material 6-Iodouridine
CAS 105967-11-1
Formula Weight 370.1
MDL Number MFCD21607789
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66634MD

 
alfa aesar™
H66634MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 370.1
Chemical Name or Material 6-Iodouridine
Physical Form Powder
Assay 96%
MDL Number MFCD21607789
CAS 105967-11-1
Density 2.27g/mL
Color Pale yellow
Quantity 250mg
Sensitivity Light Sensitive