Please login to your online account to display your discounted pricing

6-Iodonordihydrocapsaicin, Tocris

Potent, competitive vanilloid receptor antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material 6-Iodonordihydrocapsaicin
Assay Percent Range >99%
Molecular Formula C17H26INO3
Formula Weight 419.3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

197550

 
r & d systems
RDS197550
50mg Each for $844.73

197510

 
r & d systems
RDS197510
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 419.3
Quantity 50mg
Chemical Name or Material 6-Iodonordihydrocapsaicin
CAS 859171-97-4
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Molecular Formula C17H26INO3
Safety and Handling

Store at Room Temperature