Please login to your online account to display your discounted pricing

6-Iodo-2-methyl-1,3-benzothiazole, 90%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC54711DA

 View more versions of this product

Catalog No. CC54711DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 275.11
Quantity 1g
Assay 90%
Packaging Amber Glass Bottle
Assay Percent Range 90%
Molecular Formula C8H6INS
CAS 68867-20-9
Chemical Name or Material 6-Iodo-2-methyl-1, 3-benzothiazole

Specifications

Chemical Name or Material 6-Iodo-2-methyl-1, 3-benzothiazole
Assay 90%
Assay Percent Range 90%
Molecular Formula C8H6INS
Formula Weight 275.11