Please login to your online account to display your discounted pricing

6-Iodo-2-methyl-1,3-benzothiazole, 90%, Maybridge

Chemical Identifiers

Chemical Name or Material 6-Iodo-2-methyl-1, 3-benzothiazole
Assay 90%
Assay Percent Range 90%
Molecular Formula C8H6INS
Formula Weight 275.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC54711DA

  SDS
fisher scientific company
CC54711DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC54711DE

 
fisher scientific company
CC54711DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC54711CB

  SDS
fisher scientific company
CC54711CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 275.11
Quantity 1g
Assay 90%
Molecular Formula C8H6INS
CAS 68867-20-9
Assay Percent Range 90%
Packaging Amber Glass Bottle
Chemical Name or Material 6-Iodo-2-methyl-1, 3-benzothiazole