missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Iodo-2,3-dihydro-1,4-benzodioxine, 97%, Maybridge™

Chemical Identifiers

CAS 57744-67-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
MDL Number MFCD00221460
InChI Key HZTMYTXWFHBHDC-UHFFFAOYSA-N
Synonym 1,4-benzodioxin,2,3-dihydro-6-iodo, 2,3-dihydro-6-iodobenzo 1,4 dioxin, 3,4-ethylenedioxyiodobenzene, 6-iodo-1,4-benzodioxane, 6-iodo-2,3-dihydrobenzo b 1,4 dioxine, 6-iodo-2h,3h-benzo e 1,4-dioxin, 6-iodobenzodioxane, acmc-1azse
PubChem CID 2776176
IUPAC Name 6-iodo-2,3-dihydro-1,4-benzodioxine
SMILES IC1=CC=C2OCCOC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC01311DA
SDS View Documents
Fisher Scientific Company
CC01311DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

57744-67-9
262.05
HZTMYTXWFHBHDC-UHFFFAOYSA-N
2776176
IC1=CC=C2OCCOC2=C1
C8H7IO2
MFCD00221460
1,4-benzodioxin,2,3-dihydro-6-iodo, 2,3-dihydro-6-iodobenzo 1,4 dioxin, 3,4-ethylenedioxyiodobenzene, 6-iodo-1,4-benzodioxane, 6-iodo-2,3-dihydrobenzo b 1,4 dioxine, 6-iodo-2h,3h-benzo e 1,4-dioxin, 6-iodobenzodioxane, acmc-1azse
6-iodo-2,3-dihydro-1,4-benzodioxine

Specifications

Amber Glass Bottle
57744-67-9
100.0
C8H7IO2
1,4-benzodioxin,2,3-dihydro-6-iodo, 2,3-dihydro-6-iodobenzo 1,4 dioxin, 3,4-ethylenedioxyiodobenzene, 6-iodo-1,4-benzodioxane, 6-iodo-2,3-dihydrobenzo b 1,4 dioxine, 6-iodo-2h,3h-benzo e 1,4-dioxin, 6-iodobenzodioxane, acmc-1azse
IC1=CC=C2OCCOC2=C1
262.05
262.04
6-Iodo-2, 3-dihydro-1, 4-benzodioxine
1g
96.5
97%
MFCD00221460
HZTMYTXWFHBHDC-UHFFFAOYSA-N
6-iodo-2,3-dihydro-1,4-benzodioxine
2776176
97%
SDS
Product Certifications