Please login to your online account to display your discounted pricing

6-Iodo-2,3-dihydro-1,4-benzodioxine, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC01311EA

 View more versions of this product

Catalog No. CC01311EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 262.04
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 6-Iodo-2, 3-dihydro-1, 4-benzodioxine
MDL Number MFCD00221460
CAS 57744-67-9
CAS Min % 96.5
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C8H7IO2

Specifications

Chemical Name or Material 6-Iodo-2, 3-dihydro-1, 4-benzodioxine
CAS 57744-67-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7IO2
Formula Weight 262.04
MDL Number MFCD00221460